Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025466.1 | NP_079742.1 | 4, 5 | 357 | ||
GenBank mRNA | AK008622.1 | - | 4, 5 | 357 | ||
AK008640.1 | - | 4, 5 | 355 | |||
AK008641.1 | - | 4, 5 | 357 | |||
AK008647.1 | - | 4, 5 | 354 | |||
AK008722.1 | - | 4, 5 | 355 | |||
AK008745.1 | - | 4, 5 | 357 | |||
AK008933.1 | - | 4, 5 | 359 | |||
AK008956.1 | - | 4, 5 | 354 | |||
AK008990.1 | - | 4, 5 | 359 | |||
AK009145.1 | - | 4, 5 | 355 | |||
AK019050.1 | - | 4, 5 | 358 | |||
AK028136.1 | - | 4, 5 | 353 | |||
AY456960.1 | - | 4, 5 | 357 | |||
BC099446.1 | - | 4, 5 | 340 | 30521253 | ||
BC132540.1 | - | 4, 5 | 347 | 40130817 | ||
BC132542.1 | - | 4, 5 | 347 | 40130819 |