Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810032O08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007675.1 | - | 5 | 530 | ||
AK013681.1 | - | 4 | 329 | |||
AK037979.1 | - | 5 | 521 | |||
AK088915.1 | - | 5 | 512 | |||
BC055707.1 | - | 5 | 515 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_025472.1 | - | 521 | Chr. - - [-] on Build |