Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291197.1 | NP_001278126.1 | 3, 4 | 483 | ||
NM_001291200.1 | NP_001278129.1 | 3, 4 | 483 | |||
NM_025482.3 | NP_079758.2 | 3, 4 | 483 | |||
GenBank mRNA | AK013085.1 | - | 3, 4 | 425 | ||
AK013188.1 | - | 3, 4 | 436 | |||
AK147885.1 | - | 3, 4 | 443 | |||
AK159925.1 | - | 3, 4 | 411 | |||
AK171825.1 | - | 3, 4 | 445 | |||
BC006886.1 | - | 3, 4 | 372 | 2647702 | ||
CN721218.1 | - | 2, 3 | 337 | |||
DV047279.1 | - | 3, 4 | 469 |