Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025486.2 | NP_079762.1 | 5 | 422 | ||
GenBank mRNA | AK002499.1 | - | 5 | 410 | ||
AK005662.1 | - | 5 | 406 | |||
BC005665.1 | - | 5 | 377 | |||
BC024608.1 | - | 5 | 373 | 4242513 |