Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5sl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290487.1 | NP_001277416.1 | 2, 3 | 243 | ||
NM_025504.4 | NP_079780.2 | 2, 3 | 1168 | |||
GenBank mRNA | AK002370.1 | - | 2, 3 | 223 | ||
AK009150.1 | - | 2, 3 | 226 | |||
AK039669.1 | - | 2, 3 | 1169 | |||
BC024867.1 | - | 1, 2 | 199 | 4951552 |