Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160208.1 | NP_001153680.1 | 4 | 600 | ||
NM_001160209.1 | NP_001153681.1 | 3 | 687 | |||
NM_025505.4 | NP_079781.2 | 5 | 621 | |||
GenBank mRNA | AK005489.1 | - | 3 | 421 | ||
AK006544.1 | - | 4 | 604 | |||
AK029787.1 | - | 5 | 622 | |||
AK167493.1 | - | 3 | 694 | |||
AK168933.1 | - | 3 | 450 | |||
AK171167.1 | - | 3 | 479 | |||
BC027025.1 | - | 3 | 462 | 5374749 |