Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031113.1 | - | 2426 | |||
AK090053.1 | - | 2383 | ||||
AK134827.1 | - | 2338 | ||||
AK134969.1 | - | 2338 | ||||
AK136814.1 | - | 976 | ||||
AK161887.1 | - | 335 | ||||
AK165574.1 | - | 2380 |