Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000152.4 | NP_000143.2 | 8 | 1640 | ||
NM_001079803.2 | NP_001073271.1 | 9 | 1455 | |||
NM_001079804.2 | NP_001073272.1 | 8 | 1375 | |||
XM_005257193.2 | XP_005257250.1 | 9 | 1441 | |||
XM_005257194.4 | XP_005257251.1 | 9 | 1481 | |||
GenBank mRNA | AK299462.1 | - | 1419 | |||
BC040431.1 | - | 9 | 1387 | 5739991 | ||
DQ907243.1 | - | 7 | 1243 | |||
M34424.1 | - | 7 | 1275 | |||
Y00839.1 | - | 8 | 1462 |