Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291197.1 | NP_001278126.1 | 8 | 1324 | ||
NM_001291200.1 | NP_001278129.1 | 7 | 1279 | |||
NM_001291201.1 | NP_001278130.1 | 7 | 1264 | |||
NM_001291202.1 | NP_001278131.1 | 7 | 1261 | |||
NM_001291203.1 | NP_001278132.1 | 6 | 1219 | |||
NM_001291204.1 | NP_001278133.1 | 5 | 1070 | |||
NM_025482.3 | NP_079758.2 | 8 | 1321 | |||
GenBank mRNA | AK013085.1 | - | 8 | 1257 | ||
AK013188.1 | - | 8 | 1274 | |||
AK051268.1 | - | 7 | 1205 | |||
AK147885.1 | - | 7 | 1239 | |||
AK155987.1 | - | 6 | 1156 | |||
AK161092.1 | - | 6 | 1175 | |||
BC006886.1 | - | 8 | 1210 | 2647702 |