Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025529.3 | NP_079805.1 | 3, 4 | 487 | ||
GenBank mRNA | AK002605.1 | - | 3, 4 | 425 | ||
AK009700.1 | - | 3, 4 | 425 | |||
AK045197.1 | - | 3, 4 | 488 | |||
AK170076.1 | - | 3, 4 | 427 | |||
BC056443.1 | - | 3, 4 | 427 | 6822891 |