Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025535.2 | NP_079811.1 | 6 | 591 | ||
GenBank mRNA | AK002327.1 | - | 6 | 568 | ||
AK010187.1 | - | 6 | 574 | |||
AK013559.1 | - | 6 | 553 | |||
AK150906.1 | - | 6 | 547 | |||
AK155429.1 | - | 6 | 561 | |||
BC082550.1 | - | 6 | 560 | 30357800 | ||
BC085178.1 | - | 6 | 544 | 30701201 |