Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025542.2 | NP_079818.1 | 9 | 1358 | ||
GenBank mRNA | AK028792.1 | - | 9 | 1347 | ||
AK053258.1 | - | 9 | 1371 | |||
AK077823.1 | - | 9 | 1359 | |||
AK170141.1 | - | 9 | 1335 | |||
AK182599.1 | - | 1 | 332 | |||
BC049072.1 | - | 9 | 1351 | 5701585 | ||
BC096692.1 | - | 9 | 1306 | 5255425 |