Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025607.3 | NP_079883.3 | 3 | 318 | ||
GenBank mRNA | AK010854.1 | - | 3 | 331 | ||
AK014094.1 | - | 2 | 127 | |||
AK018088.1 | - | 1 | 945 | |||
AK018849.1 | - | 4 | 439 | |||
AK159788.1 | - | 3 | 331 | |||
AK166904.1 | - | 3 | 330 | |||
AK166995.1 | - | 3 | 331 | |||
AK167396.1 | - | 3 | 331 | |||
AK167633.1 | - | 3 | 331 | |||
AK167935.1 | - | 3 | 331 | |||
AK168670.1 | - | 3 | 332 | |||
AK178203.1 | - | 1 | 13 | |||
BC005559.1 | - | 3 | 342 | |||
BC083119.1 | - | 3 | 319 | 5374213 |