Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025614.3 | NP_079890.1 | 2 | 306 | ||
GenBank mRNA | AF503942.1 | - | 2 | 198 | ||
AK003262.2 | - | 2 | 260 | |||
AK005278.1 | - | 2 | 218 | |||
AK009510.1 | - | 2 | 175 | |||
AK011277.1 | - | 2 | 306 | |||
AK012504.1 | - | 2 | 277 | |||
BC062136.1 | - | 2 | 188 | 6508909 |