Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwint | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293683.1 | NP_001280612.1 | 2 | 153 | ||
NM_001293684.1 | NP_001280613.1 | 2 | 153 | |||
NM_025635.4 | NP_079911.1 | 2 | 153 | |||
GenBank mRNA | AK004305.1 | - | 2 | 105 | ||
AK008144.1 | - | 2 | 103 | |||
AK013556.1 | - | 2 | 88 | |||
AK075977.1 | - | 2 | 99 | |||
AK082215.1 | - | 2 | 103 | |||
AK134380.1 | - | 2 | 105 | |||
AK184469.1 | - | 2 | 52 | |||
BC013559.1 | - | 2 | 71 | |||
BC027100.1 | - | 2 | 110 | 5359154 | ||
BC034870.1 | - | 2 | 82 | 1382192 | ||
BY240779.1 | - | 2 | 154 |