Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080158.1 | NP_001073627.1 | 5 | 609 | ||
NM_025639.4 | NP_079915.1 | 6 | 610 | |||
NM_178269.3 | NP_840000.2 | 5 | 518 | |||
GenBank mRNA | AK010885.1 | - | 6 | 602 | ||
AK011191.1 | - | 6 | 587 | |||
AK011227.1 | - | 6 | 594 | |||
AK011464.1 | - | 6 | 611 | |||
AK011755.1 | - | 6 | 603 | |||
AK011793.1 | - | 6 | 597 | |||
AK017679.1 | - | 6 | 603 | |||
AK168203.1 | - | 5 | 499 | |||
AY243536.1 | - | 6 | 571 | |||
AY243537.1 | - | 6 | 571 | |||
BC009160.1 | - | 6 | 592 | 2655787 | ||
BY710734.1 | - | 6 | 610 |