Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsfm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025537.3 | NP_079813.1 | 3 | 286 | ||
GenBank mRNA | AJ277891.1 | - | 2 | 123 | ||
AK009898.1 | - | 3 | 306 | |||
AK012240.1 | - | 3 | 277 | |||
AK018591.1 | - | 3 | 275 | |||
AK020437.1 | - | 3 | 271 | |||
AK141183.1 | - | 3 | 276 | |||
AK169164.1 | - | 3 | 277 | |||
AK171656.1 | - | 3 | 283 | |||
AK172112.1 | - | 3 | 283 | |||
BC034286.1 | - | 3 | 265 | |||
BC057904.1 | - | 3 | 286 | 5012380 |