Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025544.2 | NP_079820.2 | 5 | 513 | ||
GenBank mRNA | AB049947.1 | - | 5 | 389 | ||
AK003126.1 | - | 5 | 434 | |||
AK005131.2 | - | 5 | 464 | |||
AK008623.1 | - | 5 | 465 | |||
AK010330.1 | - | 5 | 476 | |||
AK050396.1 | - | 5 | 517 | |||
AK140877.1 | - | 5 | 518 | |||
BC029193.1 | - | 5 | 432 | 4457237 | ||
BC055861.1 | - | 5 | 412 | 3587953 |