Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025547.3 | NP_079823.2 | 2 | 445 | ||
GenBank mRNA | AK002299.1 | - | 2 | 421 | ||
AK010525.1 | - | 2 | 440 | |||
AK011934.1 | - | 2 | 433 | |||
AK034645.1 | - | 2 | 412 | |||
BC025173.1 | - | 2 | 414 | 2655114 | ||
BC085282.1 | - | 2 | 415 | 30467998 |