Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000029.3 | NP_000020.1 | 2, 3 | 1372 | ||
GenBank mRNA | AB209769.1 | - | 2, 3 | 884 | ||
AK222798.1 | - | 2, 3 | 903 | |||
AK222815.1 | - | 2, 3 | 904 | |||
AK293507.1 | - | 3, 4 | 657 | |||
AK307978.1 | - | 3, 4 | 818 | |||
AK312291.1 | - | 2, 3 | 982 | |||
BC011519.1 | - | 2, 3 | 892 | 4213559 | ||
BT006851.1 | - | 1, 2 | 864 | |||
K02215.1 | - | 2, 3 | 903 | |||
M69110.1 | - | 1, 2 | 878 |