Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025575.3 | NP_079851.2 | 4 | 851 | ||
GenBank mRNA | AK003071.1 | - | 4 | 844 | ||
AK005249.1 | - | 4 | 754 | |||
AK005510.1 | - | 4 | 788 | |||
AK007882.1 | - | 4 | 850 | |||
AK009684.1 | - | 4 | 815 | |||
AK011753.1 | - | 4 | 847 | |||
AK013220.1 | - | 4 | 824 | |||
AK017524.1 | - | 3 | 683 | |||
AK032098.1 | - | 3 | 686 | |||
AK075770.1 | - | 4 | 816 | |||
AK148305.1 | - | 4 | 789 | |||
AK210170.1 | - | 1 | 205 | |||
AK216375.1 | - | 1 | 209 | |||
AK219257.1 | - | 1 | 209 | |||
BC053943.1 | - | 3 | 685 | |||
BC061084.1 | - | 3 | 725 | 6596164 |