Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164355.1 | NP_001157827.1 | 6 | 739 | ||
NM_025581.4 | NP_079857.3 | 6 | 803 | |||
GenBank mRNA | AK006442.1 | - | 6 | 700 | ||
AK007138.1 | - | 5 | 571 | |||
AK013233.1 | - | 6 | 701 | |||
AK021226.1 | - | 6 | 703 | |||
AK172500.1 | - | 6 | 706 | |||
BC019940.1 | - | 6 | 654 | 3709649 | ||
DQ926863.1 | - | 6 | 681 | |||
DQ926864.1 | - | 6 | 745 |