Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025599.3 | NP_079875.2 | 4 | 419 | ||
GenBank mRNA | AK012168.1 | - | 4 | 400 | ||
AK019979.1 | - | 4 | 395 | |||
BC038891.1 | - | 4 | 306 | 3371329 |