Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025607.3 | NP_079883.3 | 5 | 999 | ||
GenBank mRNA | AK010854.1 | - | 5 | 1012 | ||
AK014094.1 | - | 4 | 808 | |||
AK159788.1 | - | 5 | 1012 | |||
AK166904.1 | - | 5 | 1011 | |||
AK166995.1 | - | 5 | 1012 | |||
AK167396.1 | - | 5 | 1012 | |||
AK167633.1 | - | 5 | 1012 | |||
AK167935.1 | - | 5 | 1012 | |||
AK168670.1 | - | 5 | 1013 | |||
AK207916.1 | - | 1 | 143 | |||
AK208815.1 | - | 1 | 296 | |||
AK218504.1 | - | 1 | 143 | |||
BC005559.1 | - | 5 | 1023 | |||
BC083119.1 | - | 5 | 1000 | 5374213 |