Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025535.2 | NP_079811.1 | 6, 7 | 634 | ||
GenBank mRNA | AK002327.1 | - | 6, 7 | 611 | ||
AK010187.1 | - | 6, 7 | 617 | |||
AK013559.1 | - | 6, 7 | 596 | |||
AK150906.1 | - | 6, 7 | 590 | |||
AK155429.1 | - | 6, 7 | 604 | |||
BC082550.1 | - | 6, 7 | 603 | 30357800 | ||
BC085178.1 | - | 6, 7 | 587 | 30701201 |