Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025558.5 | NP_079834.2 | 5 | 3255 | ||
GenBank mRNA | AK052810.1 | - | 5 | 3255 | ||
AK088970.1 | - | 5 | 3255 | |||
AK165543.1 | - | 5 | 3257 | |||
AK168422.1 | - | 5 | 3245 | |||
BC046479.1 | - | 5 | 3218 | |||
BC046810.1 | - | 5 | 3221 | |||
BC048147.1 | - | 5 | 3245 | |||
BC054749.1 | - | 5 | 3276 | 6410090 | ||
BC058812.1 | - | 5 | 3216 | 6530835 | ||
BC062980.1 | - | 5 | 3269 | 6842939 |