Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magohb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025564.2 | NP_079840.2 | 5 | 577 | ||
GenBank mRNA | AK006303.1 | - | 5 | 567 | ||
AK007769.1 | - | 5 | 564 | |||
AK008200.1 | - | 5 | 579 | |||
AK013310.1 | - | 5 | 562 | |||
BC027505.1 | - | 5 | 550 | 3372083 |