Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pam16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025571.1 | NP_079847.1 | 4, 5 | 392 | ||
GenBank mRNA | AB073618.1 | - | 4, 5 | 365 | ||
AF349454.1 | - | 4, 5 | 377 | |||
AK003227.1 | - | 4, 5 | 382 | |||
AK008380.1 | - | 4, 5 | 392 | |||
BC024346.1 | - | 4, 5 | 372 | 4989038 | ||
BC096419.1 | - | 4, 5 | 381 | 1546951 |