Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025590.4 | NP_079866.2 | 15, 16 | 1752 | ||
GenBank mRNA | AF416923.1 | - | 15, 16 | 1690 | ||
AK031588.1 | - | 15, 16 | 1669 | |||
AK038546.1 | - | 15, 16 | 1690 | |||
AK048670.1 | - | 15, 16 | 1752 | |||
AK052332.1 | - | 15, 16 | 1768 | |||
AK122350.1 | - | 14, 15 | 1539 | |||
AK170189.1 | - | 8, 9 | 775 | |||
AK171255.1 | - | 8, 9 | 776 | |||
BC042492.1 | - | 15, 16 | 1687 | 4240966 |