Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000048.3 | NP_000039.2 | 3 | 290 | ||
NM_001024943.1 | NP_001020114.1 | 2 | 414 | |||
NM_001024944.1 | NP_001020115.1 | 2 | 414 | |||
NM_001024946.1 | NP_001020117.1 | 2 | 414 | |||
GenBank mRNA | AK300519.1 | - | 3 | 245 | ||
AK310912.1 | - | 2 | 420 | |||
AK316334.1 | - | 2 | 310 | |||
AY203938.1 | - | 2 | 1289 | |||
BC008195.1 | - | 3 | 252 | 3048146 | ||
BC033146.1 | - | 2 | 225 | 4101398 | ||
BQ949510.1 | - | 2 | 414 | |||
BX328836.2 | - | 3 | 310 | |||
J03058.1 | - | 3 | 248 | |||
KU177920.1 | - | 1 | 179 | |||
KU177921.1 | - | 1 | 179 | |||
KU177922.1 | - | 1 | 179 | |||
M14218.1 | - | 2 | 293 | |||
M57638.1 | - | 2 | 224 | |||
Y00753.1 | - | 2 | 224 |