Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025614.3 | NP_079890.1 | 5 | 715 | ||
GenBank mRNA | AF503942.1 | - | 5 | 607 | ||
AK003262.2 | - | 5 | 669 | |||
AK005278.1 | - | 5 | 627 | |||
AK009510.1 | - | 5 | 584 | |||
AK011277.1 | - | 5 | 715 | |||
BC062136.1 | - | 5 | 597 | 6508909 |