Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080974.2 | NP_001074443.1 | 7 | 599 | ||
NM_025618.3 | NP_079894.2 | 7 | 639 | |||
GenBank mRNA | AK008404.1 | - | 7 | 591 | ||
AK008783.1 | - | 7 | 640 | |||
AK008970.1 | - | 7 | 588 | |||
AK134747.1 | - | 7 | 601 | |||
AK146071.1 | - | 7 | 596 | |||
AK146911.1 | - | 7 | 580 | |||
AK168572.1 | - | 7 | 580 | |||
AK168792.1 | - | 7 | 580 | |||
AK168900.1 | - | 7 | 571 | |||
BC065790.1 | - | 7 | 566 | 948616 | ||
BY708689.1 | - | 7 | 636 |