Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025633.3 | NP_079909.1 | 3 | 296 | ||
GenBank mRNA | AK005241.1 | - | 3 | 293 | ||
AK010067.1 | - | 3 | 296 |