Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025641.3 | NP_079917.1 | 4 | 421 | ||
GenBank mRNA | AK003266.1 | - | 4 | 386 | ||
AK003539.1 | - | 4 | 364 | |||
AK008953.1 | - | 163 | ||||
AK011734.1 | - | 783 | ||||
AK012552.1 | - | 4 | 399 | |||
AK018712.1 | - | 4 | 399 | |||
AK019085.1 | - | 4 | 278 | |||
AK028137.1 | - | 2 | 163 | |||
AK054005.1 | - | 4 | 387 | |||
AK088185.1 | - | 4 | 386 | |||
BC011388.1 | - | 4 | 365 | 3597862 | ||
BY702808.1 | - | 4 | 394 |