Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025664.5 | NP_079940.2 | 2 | 236 | ||
GenBank mRNA | AK139802.1 | - | 2 | 270 | ||
AK149739.1 | - | 2 | 250 | |||
AK149849.1 | - | 2 | 290 | |||
AK155830.1 | - | 271 | ||||
AK157517.1 | - | 2 | 252 | |||
AK170339.1 | - | 252 | ||||
AK202748.1 | - | 2 | 85 | |||
AK202820.1 | - | 2 | 85 | |||
AK208099.1 | - | 1 | 76 | |||
AK208568.1 | - | 2 | 85 | |||
AK218282.1 | - | 2 | 85 | |||
BC014814.1 | - | 2 | 236 | 3257422 |