Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025665.2 | NP_079941.1 | 2, 3 | 174 | ||
GenBank mRNA | AK011937.1 | - | 2, 3 | 175 | ||
AK045231.1 | - | 2, 3 | 187 | |||
AK160190.1 | - | 2, 3 | 184 | |||
BC027564.1 | - | 2, 3 | 175 | 3468252 |