Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281726.1 | NP_001268655.1 | 2, 3 | 423 | ||
XM_006713314.3 | XP_006713377.1 | 3, 4 | 703 | |||
XM_011534041.2 | XP_011532343.1 | 3, 4 | 540 | |||
XM_017007088.1 | XP_016862577.1 | 4, 5 | 816 | |||
GenBank mRNA | AK294301.1 | - | 4, 5 | 555 | ||
AK297033.1 | - | 2, 3 | 415 | |||
AK301838.1 | - | 2, 3 | 615 | |||
AK313252.1 | - | 2, 3 | 406 | |||
BC012099.1 | - | 2, 3 | 349 | 4559618 | ||
BE814688.1 | - | 2, 3 | 446 | |||
BM839350.1 | - | 4, 5 | 506 | |||
BM919775.1 | - | 2, 3 | 416 | |||
BP326502.1 | - | 2, 3 | 423 | |||
U03274.1 | - | 2, 3 | 352 |