Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000063.5 | NP_000054.2 | 8 | 1379 | ||
NM_001145903.2 | NP_001139375.1 | 6 | 983 | |||
NM_001178063.2 | NP_001171534.1 | 4 | 529 | |||
NM_001282457.1 | NP_001269386.1 | 4 | 586 | |||
NM_001282458.1 | NP_001269387.1 | 8 | 1407 | |||
GenBank mRNA | AK096258.1 | - | 5 | 685 | ||
AK222537.1 | - | 8 | 1131 | |||
AK222783.1 | - | 8 | 1103 | |||
AK298311.1 | - | 4 | 529 | |||
AK298808.1 | - | 4 | 586 | |||
AK300892.1 | - | 6 | 743 | |||
AK300930.1 | - | 8 | 1145 | |||
AK307930.1 | - | 6 | 739 | |||
AK312581.1 | - | 8 | 1213 | |||
BC043484.1 | - | 8 | 1114 | 5763774 | ||
BX537504.1 | - | 2 | 1116 | |||
X04481.1 | - | 8 | 1139 |