Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025718.3 | NP_079994.2 | 4 | 413 | ||
GenBank mRNA | AK003619.1 | - | 4 | 414 | ||
AK014633.1 | - | 3 | 605 |