Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933417A18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025750.3 | NP_080026.2 | 1, 2 | 298 | ||
GenBank mRNA | AK016778.1 | - | 1, 2 | 263 | ||
AK016838.1 | - | 1, 2 | 299 | |||
BC100450.1 | - | 1, 2 | 288 | 30919421 | ||
BC145747.1 | - | 1, 2 | 253 | 40131050 |