Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025650.2 | NP_079926.1 | 3 | 371 | ||
GenBank mRNA | AK003033.1 | - | 3 | 381 | ||
AK003443.1 | - | 3 | 351 | |||
AK013513.1 | - | 3 | 319 | |||
AK146292.1 | - | 3 | 513 | |||
AK167946.1 | - | 3 | 367 | |||
AK168974.1 | - | 3 | 336 | |||
BC034408.1 | - | 3 | 335 | 4235605 | ||
BU529273.1 | - | 3 | 372 |