Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163464.1 | NP_001156936.1 | 3 | 268 | ||
NM_025659.4 | NP_079935.1 | 3 | 619 | |||
GenBank mRNA | AK008928.1 | - | 3 | 341 | ||
AK018381.1 | - | 3 | 340 | |||
AK033965.1 | - | 3 | 340 | |||
AK037144.1 | - | 3 | 269 | |||
AK172098.1 | - | 3 | 338 | |||
BC058260.1 | - | 3 | 329 | 6474610 |