Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025665.2 | NP_079941.1 | 3, 4 | 289 | ||
GenBank mRNA | AK011937.1 | - | 3, 4 | 290 | ||
AK045231.1 | - | 3, 4 | 302 | |||
AK160190.1 | - | 3, 4 | 299 | |||
BC027564.1 | - | 3, 4 | 290 | 3468252 |