Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025689.4 | NP_079965.2 | 2, 3 | 391 | ||
GenBank mRNA | AK014056.1 | - | 1, 2 | 331 | ||
AK017860.1 | - | 2, 3 | 388 | |||
AK137013.1 | - | 2, 3 | 395 | |||
AK141240.1 | - | 2, 3 | 394 | |||
BC098221.1 | - | 1, 2 | 337 | 949536 | ||
BC116786.1 | - | 1, 2 | 249 | 40126105 | ||
BC116788.1 | - | 1, 2 | 249 | 40126107 |