Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spryd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025697.3 | NP_079973.1 | 3, 4 | 646 | ||
GenBank mRNA | AK018044.1 | - | 3, 4 | 524 | ||
AK018153.1 | - | 3, 4 | 647 | |||
AK018290.1 | - | 3, 4 | 496 | |||
AK137685.1 | - | 495 | ||||
AK167306.1 | - | 3, 4 | 488 | |||
AK169059.1 | - | 3, 4 | 495 | |||
BC037015.1 | - | 3, 4 | 480 | 5344409 |