Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172481.1 | NP_001165952.1 | 4 | 729 | ||
NM_025711.3 | NP_079987.2 | 4 | 746 | |||
GenBank mRNA | AF316825.1 | - | 3 | 466 | ||
AK014504.1 | - | 4 | 702 | |||
AK085920.1 | - | 4 | 701 | |||
BC034888.1 | - | 4 | 648 | 1365428 | ||
BC145904.1 | - | 4 | 644 | 40131207 | ||
BC145930.1 | - | 4 | 627 | 40131233 |