Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4921530L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025733.2 | NP_080009.1 | 2 | 494 | ||
GenBank mRNA | AK005846.1 | - | 2 | 464 | ||
AK014987.1 | - | 2 | 495 | |||
BC049573.1 | - | 2 | 448 | 6707442 | ||
EU477368.1 | - | 1 | 305 |