Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000091.4 | NP_000082.2 | 3 | 360 | ||
XM_005246277.3 | XP_005246334.1 | 3 | 336 | |||
XM_005246280.3 | XP_005246337.1 | 3 | 336 | |||
XM_006712245.3 | XP_006712308.1 | 3 | 336 | |||
XM_011510555.1 | XP_011508857.1 | 3 | 336 | |||
XM_017003295.1 | XP_016858784.1 | 3 | 336 | |||
GenBank mRNA | X80031.1 | - | 3 | 359 |