Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025666.2 | NP_079942.1 | 11 | 2832 | ||
GenBank mRNA | AK088208.1 | - | 11 | 2834 | ||
AK149989.1 | - | 11 | 2787 | |||
BC051678.1 | - | 8 | 2360 | |||
BC058535.1 | - | 10 | 2516 | |||
BC079531.1 | - | 10 | 2529 | |||
BC094323.1 | - | 11 | 2839 | |||
BC094423.1 | - | 11 | 2790 | 6812389 |