Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025668.3 | NP_079944.1 | 4, 5 | 508 | ||
GenBank mRNA | AK014310.1 | - | 4, 5 | 522 | ||
AK017504.1 | - | 4, 5 | 513 | |||
AK077510.1 | - | 4, 5 | 518 | |||
AK134578.1 | - | 4, 5 | 509 | |||
AK168173.1 | - | 4, 5 | 513 | |||
AK211106.1 | - | 31 | ||||
AK212862.1 | - | 31 | ||||
BC010547.1 | - | 3, 4 | 326 | |||
BC050150.1 | - | 4, 5 | 518 | 30026746 |